Learn More
Lonza™ BioWhittaker™ PowerCHO™ CD Medium
Support the growth of CHO K1, DXB11, DG44 and GS systems
Brand:  Lonza™ BE12-771Q
Additional Details : Weight : 1.00000kg
 View more versions of this product
Product Code. 11615260
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Description
Description
PowerCHO 1, 2, and 3 Chemically Defined, Serum-free CHO Media are the next generation in CHO media, optimized for both cell growth and protein production. For therapeutic bioprocessing applications, these protein-free formulations also facilitate both downstream purification and regulatory compliance.
  • Non-animal origin media
  • Hydrolysate-free, serum-free
  • Contains HEPES and Pluronic™ F-68
  • Free of glutamine, hypoxanthine, thymidine and phenol red
  • Maintain high viability (>90%) at high cell densities
  • Support high-density, large scale production volumes through balanced formulation
Specifications
Specifications
2° to 8°C
CHO
No Glutamine, No Phenol Red, No Hypoxanthine, No Thymidine
1 L
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Promotions
Promotions