Learn More
Lonza™ BioWhittaker™ Dulbecco's Phosphate Buffered Saline (1X) w/o Calcium and Magnesium
Used to provide buffer system to maintain cell culture media in the physiological pH range of 7 to 7.6.
Brand: Lonza™
Additional Details : Weight : 1.36080kg
Description
- Serves as an irrigating, transporting or diluting fluid while maintaining cell tonicity and viability for a limited period of time
- Manufactured in accordance with cGMP regulations and are cell culture tested to assure compatibility
Specifications
15° to 30°C | |
No Calcium, No Magnesium | |
500 mL |
Dulbecco's Phosphate Buffered Saline | |
7 to 7.6 |