Få mere at vide
R&D Systems™ Biotinylated Alkyne
Beskrivelse
Indeholder
- Highly specific to the azido group.
- Can be linked to the azido group through click chemistry.
- Versatile for detection through the biotin moiety.
Provided in 20mM Tris, pH 8.0
Tekniske data
Tekniske data
| Mængde | 500 μg |
| Molekylær formel | C19H31N3O5S |
| Formel vægt | 413.53 Da |