Learn More
Thermo Scientific™ Biotinyl-[Glu1]-Gastrin I, Human
For cellular and molecular biology applications
Brand:  Thermo Scientific™ J66203.|D
Additional Details : Weight : 0.00001kg
 View more versions of this product
Product Code. 15477469
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Description
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
Specifications
Human
-30°C to -10°C
2342.60
0.5 mg
Biotin-Glu-Gly-Pro-Trp-Leu-Glu-Glu-Glu-Glu-Glu-Ala-Tyr-Gly-Trp-Met-Asp-Phe-NH2
HPLC
Solid
Biotinyl-[Glu1]-Gastrin I, human
RUO - research use only
Product Suggestions
Product Suggestions
Videos
SDS Documents
Special Offers
For Research Use Only