missing translation for 'onlineSavingsMsg'
Learn More

Biotinyl-[Glu1]-Gastrin I, Human, Alfa Aesar™

For cellular and molecular biology applications

Brand:  Alfa Aesar J66203.M¬

Additional Details : Weight : 0.00001kg

 View more versions of this product

Product Code. 15487469

  • 1782.45 DKK / 1mg


Estimated Shipment
26-10-2020
Add to basket

Specifications

Specifications

2342.60
Human
Solid
RUO
Biotinyl-[Glu1]-Gastrin I, human
HPLC
1mg
Biotin-Glu-Gly-Pro-Trp-Leu-Glu-Glu-Glu-Glu-Glu-Ala-Tyr-Gly-Trp-Met-Asp-Phe-NH2
-30° to -10°C
Solid
Videos
SDS Documents

For Research Use Only