Learn More
Biotin-TEG Azide, TRC
Product Code. 30134992
Change view
Click to view available options
Quantity:
10 mg
25 mg
100 mg
Unit Size:
100mg
10mg
25mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30134992 100 mg 100mg
30134545 25 mg 25mg
30008724 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30134992 Supplier Toronto Research Chemicals Supplier No. TRCB391600100MG
 Restricted Item
Forventet forsendelse: 10-12-2026
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 875770-34-6
Molecular Formula C18H32N6O5S
Molecular Weight (g/mol) 444.55
Synonym Azide-PEG3-biotin, N-[2-[2-[2-(2-Azidoethoxy)ethoxy]ethoxy]ethyl]-5-((3aS,4S,6aR)-2-oxohexahydro-1H-thieno[3,4-d]imidazol-4-yl)pentanamide, (3aS,4S,6aR)-N-[2-[2-[2-(2-Azidoethoxy)ethoxy]ethoxy]ethyl]hexahydro-2-oxo-1H-thieno[3,4-d]imidazole-4-pentanamide
IUPAC Name 5-[(3aS,4S,6aR)-2-oxo-1,3,3a,4,6,6a-hexahydrothieno[3,4-d]imidazol-4-yl]-N-[2-[2-[2-(2-azidoethoxy)ethoxy]ethoxy]ethyl]pentanamide
SMILES [N-]=[N+]=NCCOCCOCCOCCNC(CCCC[C@H]1[C@@H]2[C@@H](NC(N2)=O)CS1)=O
Specifications
Quantity 100 mg
InChI Formula InChI=1S/C18H32N6O5S/c19-24-21-6-8-28-10-12-29-11-9-27-7-5-20-16(25)4-2-1-3-15-17-14(13-30-15)22-18(26)23-17/h14-15,17H,1-13H2,(H,20,25)(H2,22,23,26)/t14-,15-,17-/m0/s1
Formula Weight 444.2155
Chemical Name or Material Biotin-TEG Azide