Få mere at vide
Biopterin-d3, TRC
Artikelnummer. 30342913
Skift visning
Mængde:
5 mg
Artikelnummer. Mængde
30342913 5 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30342913 Leverandør Toronto Research Chemicals Leverandørnr. TRCB3890225MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Biopterin-d3
CAS 1217838-71-5
Renhedsgrad noter HPLC
Molekylær formel C9 2H3 H8 N5 O3
Mængde 5 mg
Synonym rel-2-Amino-6-[(1R,2S)-1,2-dihydroxypropyl-3,3,3-d3]-4(3H)-pteridinone, DL-erythro-Biopterin-3',3',3'-D3, DL-erythro-Biopterin-d3, Biopterin-d3
InChI formel InChI=1S/C9H11N5O3/c1-3(15)6(16)4-2-11-7-5(12-4)8(17)14-9(10)13-7/h2-3,6,15-16H,1H3,(H3,10,11,13,14,17)/t3-,6-/m0/s1/i1D3
SMIL [2H]C([2H])([2H])[C@H](O)[C@H](O)c1cnc2nc(N)nc(O)c2n1
IUPAC navn (1R,2S)-1-(2-amino-4-hydroxypteridin-6-yl)-3,3,3-trideuteriopropane-1,2-diol
Molekylvægt (g/mol) 240.23
Formel vægt 240.105
Procent renhed >95