Learn More
Sartorius™ Biohit™ Pcu 2-Main Boards
Sartorius™ Biohit™ Pcu 2-Main Boards are replacement circuit boards for use with compatible eLine™ electronic multichannel pipettes.
Brand: Sartorius™ 731059
Additional Details : Weight : 0.10000kg
Description
eLINE™ electronic pipettes offers an attractive and ergonomic design together with electronic tip ejection operation to take the effort out of pipetting and reduce the risk of repetitive strain injuries (RSI) that are common in mechanical pipetting.
- Microprocessor controlled system enables maximum liquid handling performance with high levels of accuracy and precision
- Microprocessor controlled piston movements reduces the possibility of human error and thus also contamination of the pipette interior
- The simple keyboard, easy-to-read display and user-friendly operation and programming allows tasks to be performed quickly and effortlessly
- Offers a comprehensive range of liquid handling protocols typically required by laboratories
- Available in a range of pipetting volumes
Specifications
Pcu 2-Main Board | |
1200μL E-Line Multi Channel Pipette |