Få mere at vide
Corning™ BioCoat™ Kollagen IV rektangulær skråhalskulturkolbe
Kapacitet (metrisk):
70 mL
250 mL
750 mL
Kulturområde:
175 cm 2
25 cm2
75 cm 2
Pakningsstørrelse:
Pakke med 10
Pakke med 5
Beskrivelse
- BioCoat™ provides a superior solution for enhanced cell attachment and growth of a variety of primary cells and transformed cells in serum-free or serum-containing cultures
- Matrix components are applied as an even, optically clear coating
- All lots tested and found negative for bacteria and fungi; cellware is tested for the ability to promote firm attachment of rat cerebellar granule cells
For research use only.
Tekniske data
Tekniske data
| Lukningsmateriale | Phenolic |
| Kapacitet (metrisk) | 70 mL |
| Til brug med (applikation) | For adhesion, growth and differentiation of neuronal and transfected cell lines |
| Hals stil | Canted |
| Form | Rectangular |
| Overfladebelægning | Collagen IV |
| Sterilitet | Sterile |
| Lukketype | Plug Seal |
| Kulturområde | 25 cm2 |