Learn More
Nitritex™ BioClean-D™ Non-Sterile Visitor Lab Coats
Lightweight and comfortable. Nitritex™ BioClean-D™ Non-Sterile Visitor Coat is a disposable coat manufactured from antistatic, low linting and durable CleanTough™ fabric and features a mandarin collar, press stud fastenings, elasticated cuffs and three pockets.
3765.00 DKK
Specifications
Product Type | Controlled Environments Lab Coat |
---|---|
Gender | Unisex |
Material | Double Laminate Microporous Fabric |
Color | White |
Closure Type | Zipper |
Description
- Antistatic, low-linting and durable white CleanTough™ fabric
- Press studs for easy fastening
- Bound seams with single needle stitching
- Mandarin collar and Elasticated cuffs
- Antistatic EN 1149-5 and low particulating for use in non-critical environments
Specifications
Controlled Environments Lab Coat | |
Double Laminate Microporous Fabric | |
Zipper | |
Single-use | |
Puncture | |
Bound | |
(EU) 2016/425, EN 1149-5:2008, EN 530, EN 7854, EN 6530, EN 13934-1, EN 9073-4, EN 863, EN 13935-2 | |
Single Needle Stitching |
Unisex | |
White | |
Elastic | |
Non-sterile | |
3 | |
ISO 9001:2015 | |
Five pieces per sealed inner PE bag; six inner bags per lined carton (30 pieces) |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Product Title
Nitritex™ BioClean-D™ Non-Sterile Visitor Lab Coats