Learn More
Benchmark Scientific™ BioClave Research Autoclaves
Available in 8 L, 18 L, and 28 L chamber volumes, the BioClave Research Autoclaves are benchtop, stand-alone units that include mechanical and electrical safety interlocks. Operation is fully automatic, simply pick the appropriate program and press start.
Brand:  Benchmark Scientific™ B4000-28-220
Additional Details : Weight : 95.00000kg
 View more versions of this product
Product Code. 17924422
  • 104060.96 DKK / Each
Estimated Shipment: 26-07-2024
to see stock.
Description
Description
  • Designed specifically for the sterilization of laboratory research tools
  • Fully automatic operation - just press start
  • Built in water tank - no external connection needed
  • 8 L, 18 L and 28 L models
  • All models small enough for benchtop use
Specifications
Specifications
7.4 gal.
41.7 cm
16.4 in.
25 in.
Front-loading Benchtop Autoclave
121°C to 134°C
3 Shelves, Extra Door Seal
3
28 L
12.6 in.
32 cm
63.5 cm
250°F to 273°F
Digital
Non-sterile
63.5 x 63.5 x 55.9 cm