Få mere at vide
Thermo Scientific™ BioBasic™ SEC 120 Size Exclusion Chromatography HPLC Columns
Shop alle Thermo Scientific produkterBeskrivelse
Outstanding size separation of water soluble compounds.
Provide outstanding size separation of water soluble compounds with Thermo Scientific™ BioBasic™ SEC 120 Size Exclusion Chromatography HPLC Columns. BioBasic SEC columns provide high efficiency separations for a wide range of molecular weights from 100 to 10,000,000 daltons, allowing the analysis of proteins, peptides and other biomolecules.
- Covers separation of analytes over a wide molecular weight range
- Long column life and high column efficiencies
- Simple mechanism of interaction based on molecular size and shape
- Excellent for sample cleanup
- Straightforward method development, simple mobile phases
- 120Å pore size
Ordering Information:
For information on bulk quantities, please inquire.
Tekniske data
Tekniske data
| Kolonneformat | High Temperature Column |
| Diameter (metrisk) | 7.8 mm |
| Til brug med | Size Exclusion Chromatography |
| Længde (metrisk) | 150 mm |
| Maks. Tryk | 5800 psi (400 bar) |
| Partikelstørrelse | 5 μm |
| Porestørrelse | 120 Å |
| USP type | L33 |
| Stationær fase | Diol (OH) |
| Produktlinje | BioBasic |
| Vis mere |