missing translation for 'onlineSavingsMsg'
Få mere at vide

STEMCELL Technologies BIO-Acetoxime

Artikelnummer. 17170202
Encompass_Preferred
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
17170202 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 17170202 Leverandør STEMCELL Technologies Leverandørnr. 73322

Please to purchase this item. Need a web account? Register with us today!

Denne vare kan ikke returneres. Se returpolicy

WNT pathway activator; Inhibits GSK3

  • BIO-Acetoxime is an analog of the GSK3 inhibitor BIO (Catalog #72032)
  • It potently inhibits GSK3α and GSK3β (IC50 = 10 nM), and less potently inhibits cyclin-dependent kinases Cdk5/p25, Cdk2/A, and Cdk1/B (IC50 = 2.4, 4.3, and 63 μM, respectively; Meijer et al.; Polychronopoulos et al.) By inhibiting GSK3β, BIO-Acetoxime blocks β-catenin phosphorylation and degradation, thereby allowing it to activate transcription of WNT pathway-controlled genes (Meijer et al.)
  • Inhibits CD8+ effector T cell differentiation (Zhou et al.)
  • Suppresses viral gene expression and protects oral epithelial cells from herpes simplex virus-1 infection (Hsu & Hung)

Tekniske data

CAS 740841-15-0
Synonym 6-Bromoindirubin-3’-acetoxime; BIA; GSK-3 inhibitor X
Mængde 1 mg
Molekylær formel C18H12BrN3O3
Renhed ≥95%
Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.