missing translation for 'onlineSavingsMsg'
Få mere at vide
Få mere at vide
STEMCELL Technologies BIO
Beskrivelse
- BIO is an indirubin compound that is cell-permeable and is a selective and potent reversible inhibitor of GSK3α and GSK3β (IC50 = 5 nM) and functions as a WNT activator
- Enhances reprogramming of mouse fibroblasts, neural stem cells, and thymocytes to induced pluripotent stem (iPS) cells (Lluis et al.)
- Maintains self-renewal of mouse embryonic stem (ES) cells in the absence of LIF (Sato et al.)
- Maintains self-renewal of human ES cells in unconditioned medium through a SMAD 2/3 dependent mechanism (James et al.), Sato et al.)
- Induces proliferation of neonatal and adult rat cardiomyocytes (Tseng et al.)
- Promotes differentiation of cardiomyocytes from human ES and iPS cells (Lian et al.)
Tekniske data
Tekniske data
| CAS | 667463-62-9 |
| Synonym | GSK 3 IX; MLS 2052; 6-Bromoindirubin-3'-oxime |
| Mængde | 5 mg |
| Molekylær formel | C16H10BrN3O2 |
| Renhed | ≥98% |
Produkttitel
Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.
Ser du en mulighed for forbedring?