Learn More
Thermo Scientific™ BindIt™ Software, With CD only
Product Code. 15903314 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
Each
Unit Size:
Each
Product Code. Quantity Unit Size
15903314 Each Each
1 options
Product Code. 15903314 Supplier Thermo Scientific™ Supplier No. 5189009
 Restricted Item
Forventet forsendelse: 02-12-2026
Automate customized applications using Thermo Scientific™ BindIt™ Software, a versatile tool for protocol creation and modification.
Create and store KingFisher purification system protocols using Thermo Scientific™ BindIt™ Software. Once a protocol has been created, the protocol can either be transferred to the Thermo Scientific™ KingFisher™ instrument memory or executed directly from the software. All protocol steps have default parameters which can be changed according to the demands of the application.
Features:
  • Plates and reagents used are defined in the plate layout
  • Due to an increased QC demand, BindIt Software enables the user to generate a status report including the run log, plate layout and step parameters
  • Enables the KingFisher Flex to interface with liquid handling, robotics and plate stacking instruments, providing a fully automated solution and the highest possible throughput
Compatible with:
KingFisher, KingFisher mL, KingFisher Flex, KingFisher Duo Prime
TRUSTED_SUSTAINABILITY
Specifications
Description BindIt Software
Display XVGA monitor with 1024 by 768 resolution
For Use With (Equipment) KingFisher, KingFisher mL, KingFisher Flex, KingFisher Duo Prime
Includes CD only
Ports One serial or USB port available
Quantity Each
Memory 1 GB RAM
Operating System Windows XP Professional with Service Pack 3, Windows Vista Service with Pack 2, Windows 7 with Service Pack 1, and Windows 8