Få mere at vide
Tocris Bioscience™ BigLEN (mouse)
GREENER_CHOICE
GPR171 agonist
Brand:  Tocris Bioscience™ 6304/1
Encompass
Produktkode 15776399
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Se alternative produkter »
Denne vare kan ikke returneres. Se returpolitik
Beskrivelse
Beskrivelse
BigLEN (mouse) is a GPR171 agonist. ProSAAS-derived neuropeptide. Regulates food intake in mice. Inhibits the release of glutamate onto parvocellular neurons of the paraventricular nucleus in a process dependent upon activation of postsynaptic G proteins.
Tekniske data
Tekniske data
501036-69-7
Store at -20°C
1756.03
1 mg
Produktforslag
Produktforslag
Videoer
SDS
Dokumenter
Dokumenter
Certifikater
Kampagner
Kampagner