Få mere at vide
Bifonazole EP Impurity D (as Chloride), Mikromol™
Artikelnummer. 30463152
Skift visning
Mængde:
25 mg
100 mg
4 x 25 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30463152 4 x 25 mg
30052716 25 mg
30050999 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30463152 Leverandør Mikromol Leverandørnr. MM0318.0300254AMP
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
Bifonazole EP Impurity D (as Chloride), Mikromol
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 1,3-Bis[(biphenyl-4-yl)phenylmethyl]-1H-imidazolium Chloride
CAS 66600-13-3
Analyt- eller komponentnavne 1,3-Bis[(biphenyl-4-yl)phenylmethyl]-1H-imidazolium Chloride
Forsendelsestilstand Room Temperature
Molekylær formel C41 H33 N2 . Cl
Certificeringer/compliance ISO 9001
Synonym Bifonazole Imp. D (EP), 1,3-Bis[(biphenyl-4-yl)phenylmethyl]-1H-imidazolium Chloride, Bifonazole Imp. D (EP) as Chloride, Bifonazole Impurity D as Chloride
InChI formel InChI=1S/C41H33N2.ClH/c1-5-13-32(14-6-1)34-21-25-38(26-22-34)40(36-17-9-3-10-18-36)42-29-30-43(31-42)41(37-19-11-4-12-20-37)39-27-23-35(24-28-39)33-15-7-2-8-16-33;/h1-31,40-41H;1H/q+1;/p-1
Løsningstype Neat
SMIL [Cl-].c1ccc(cc1)C(c2ccc(cc2)c3ccccc3)[n+]4ccn(c4)C(c5ccccc5)c6ccc(cc6)c7ccccc7
IUPAC navn 1,3-bis[phenyl-(4-phenylphenyl)methyl]imidazol-1-ium;chloride
Molekylvægt (g/mol) 589.17
Mængde 4 x 25 mg
Formel vægt 588.2332 g/mol