missing translation for 'onlineSavingsMsg'
Få mere at vide

Bifonazole EP Impurity A, Mikromol™

Artikelnummer. 30457872
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
25 mg
100 mg
4 x 25 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30457872 4 x 25 mg
30051567 100 mg
30052103 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 30457872 Leverandør Mikromol Leverandørnr. MM0318.0100254AMP

Please to purchase this item. Need a web account? Register with us today!

Denne vare kan ikke returneres. Se returpolicy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

Bifonazole EP Impurity A, Mikromol
TRUSTED_SUSTAINABILITY

Specifications

Kemisk navn eller materiale (RS)-(Biphenyl-4-yl)phenylmethanol
CAS 7598-80-3
Analyt- eller komponentnavne (RS)-(Biphenyl-4-yl)phenylmethanol
Forsendelsestilstand Room Temperature
Molekylær formel C19 H16 O
Certificeringer/compliance ISO 17025
Synonym Bifonazole Imp. A (EP), (RS)-(Biphenyl-4-yl)phenylmethanol, Bifonazole Impurity A
InChI formel InChI=1S/C19H16O/c20-19(17-9-5-2-6-10-17)18-13-11-16(12-14-18)15-7-3-1-4-8-15/h1-14,19-20H
Løsningstype Neat
SMIL OC(c1ccccc1)c2ccc(cc2)c3ccccc3
IUPAC navn phenyl-(4-phenylphenyl)methanol
Molekylvægt (g/mol) 260.33
Mængde 4 x 25 mg
Formel vægt 260.1201 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.