Få mere at vide
Bifonazole EP Impurity A, Mikromol™
Artikelnummer. 30051567
Skift visning
Mængde:
25 mg
100 mg
4 x 25 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30051567 100 mg
30052103 25 mg
30457872 4 x 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30051567 Leverandør Mikromol Leverandørnr. MM0318.01
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
Bifonazole EP Impurity A
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale (RS)-(Biphenyl-4-yl)phenylmethanol
CAS 7598-80-3
Analyt- eller komponentnavne (RS)-(Biphenyl-4-yl)phenylmethanol
Forsendelsestilstand Room Temperature
Molekylær formel C19 H16 O
Certificeringer/compliance ISO 17025
Synonym Bifonazole Imp. A (EP), (RS)-(Biphenyl-4-yl)phenylmethanol, Bifonazole Impurity A
InChI formel InChI=1S/C19H16O/c20-19(17-9-5-2-6-10-17)18-13-11-16(12-14-18)15-7-3-1-4-8-15/h1-14,19-20H
SMIL OC(c1ccccc1)c2ccc(cc2)c3ccccc3
IUPAC navn phenyl-(4-phenylphenyl)methanol
Molekylvægt (g/mol) 260.33
Mængde 100 mg
Formel vægt 260.1201 g/mol