Få mere at vide
Bicalutamide-d4, TRC
Artikelnummer. 30334979
Skift visning
Mængde:
10 mg
Artikelnummer. Mængde
30334979 10 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30334979 Leverandør Toronto Research Chemicals Leverandørnr. TRCB38200210MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Bicalutamide-d4
CAS 1185035-71-5
Renhedsgrad noter HPLC
Molekylær formel C18H10D4F4N2O4S
Mængde 10 mg
Synonym N-[4-Cyano-3-trifluoromethyl)phenyl]- 3-[(4-fluorophenyl)sulfonyl]-2-hydroxy-2-methylpropanamide-d4, ICI-176334-d4, Casodex-d4
InChI formel InChI=1S/C18H14F4N2O4S/c1-17(26,10-29(27,28)14-6-3-12(19)4-7-14)16(25)24-13-5-2-11(9-23)15(8-13)18(20,21)22/h2-8,26H,10H2,1H3,(H,24,25)/i3D,4D,6D,7D
SMIL O=C(C(O)(C)CS(C1=C([2H])C([2H])=C(F)C([2H])=C1[2H])(=O)=O)NC2=CC=C(C#N)C(C(F)(F)F)=C2
IUPAC navn N-[4-cyano-3-(trifluoromethyl)phenyl]-2-hydroxy-2-methyl-3-(2,3,5,6-tetradeuterio-4-fluorophenyl)sulfonylpropanamide
Molekylvægt (g/mol) 434.4
Procent renhed >95