Få mere at vide
Tocris Bioscience™ BIBO 3304 trifluoroacetate
Beskrivelse
BIBO 3304 trifluoroacetate is a high affinity NPY Y1 receptor antagonist (IC50 values are 0.38 and 0.72 nM at human and rat receptors respectively) that displays > 2600-fold selectivity over Y2, Y4 and Y5 receptors. Inhibits NPY - and fasting-induced feeding in vivo following central administration. Also antagonizes anxiolytic-like effects of NPY.
Tekniske data
Tekniske data
| Kemisk navn eller materiale | N-[(1R)-1-[[[[4-[[(Aminocarbonyl)amino]methyl]phenyl]methyl]amino]carbonyl]-4-[(aminoiminomethyl)amino]butyl]-α-phenyl-benzeneacetamide ditrifluoroacetate |
| CAS | 2310085-85-7 |
| Analyseprocentområde | >98% |
| Mængde | 10 mg |
| Opløselighed | Soluble to 100mM in DMSO |
| Molekylær formel | C29H35N7O3.2CF3CO2H |
| Formel vægt | Observed MW: 757.69 |
For Research Use Only