Få mere at vide
Tocris Bioscience™ BI 9564
Potent and selective BRD9 and BRD7 inhibitor; orally active
Brand: Tocris Bioscience™ 5590/10
Denne vare kan ikke returneres.
Se returpolitik
Beskrivelse
BI 9564 is a potent and selective BRD9 and BRD7 inhibitor (Kd values are 14 and 239 nM, respectively). Exhibits selectivity for BRD9/7 over 48 other bromodomains, 324 kinases and 55 GPCRs. Inhibits proliferation of AML cell lines in vitro and reduces AML tumor xenograft growth in vivo. Cell permeable and orally active.Tekniske data
| 4-[4-[(Dimethylamino)methyl]-2,5-dimethoxyphenyl]-2-methyl-1,2-dihydro-2,7-naphthyridin-1-one | |
| 1883429-22-8 | |
| 10 mg |