Få mere at vide
Bezafibrate Isopropyl Ester, Mikromol™
Artikelnummer. 30050232
Skift visning
Mængde:
25 mg
100 mg
4 x 25 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30050232 100 mg
30052387 25 mg
30462913 4 x 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30050232 Leverandør Mikromol Leverandørnr. MM0063.08
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
Bezafibrate Isopropyl Ester
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Bezafibrate Isopropyl Ester
CAS 153758-85-1
Analyt- eller komponentnavne Bezafibrate Isopropyl Ester
Forsendelsestilstand Room Temperature
Molekylær formel C22 H26 Cl N O4
Certificeringer/compliance ISO 9001
Synonym Bezafibrate Isopropyl Ester, 2-[4-[2-[(4-chlorobenzoyl)amino]ethyl]phenoxy]-2-methyl-propanoic acid 1-methylethyl ester
InChI formel InChI=1S/C22H26ClNO4/c1-15(2)27-21(26)22(3,4)28-19-11-5-16(6-12-19)13-14-24-20(25)17-7-9-18(23)10-8-17/h5-12,15H,13-14H2,1-4H3,(H,24,25)
SMIL CC(C)OC(=O)C(C)(C)Oc1ccc(CCNC(=O)c2ccc(Cl)cc2)cc1
IUPAC navn propan-2-yl 2-[4-[2-[(4-chlorobenzoyl)amino]ethyl]phenoxy]-2-methylpropanoate
Molekylvægt (g/mol) 403.9
Mængde 100 mg
Formel vægt 403.155 g/mol