Få mere at vide
Bezafibrate-d6 (dimethyl-d6), TRC
Artikelnummer. 30334180
Skift visning
Mængde:
0.5 mg
Pakningsstørrelse:
0.5mg
Artikelnummer. Mængde unitSize
30334180 0.5 mg 0.5mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30334180 Leverandør Toronto Research Chemicals Leverandørnr. TRCB3410010.5MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Bezafibrate D6 (dimethyl D6)
CAS 1219802-74-0
Molekylær formel C19 2H6 H14 Cl N O4
Mængde 0.5 mg
Synonym Bezafibrate D6 (dimethyl D6), 2-[4-[2-[(4-Chlorobenzoyl)amino]ethyl]phenoxy]-3,3,3-trideuterio-2-(trideuteriomethyl)propanoic acid
InChI formel InChI=1S/C19H20ClNO4/c1-19(2,18(23)24)25-16-9-3-13(4-10-16)11-12-21-17(22)14-5-7-15(20)8-6-14/h3-10H,11-12H2,1-2H3,(H,21,22)(H,23,24)/i1D3,2D3
SMIL [2H]C([2H])([2H])C(Oc1ccc(CCNC(=O)c2ccc(Cl)cc2)cc1)(C(=O)O)C([2H])([2H])[2H]
IUPAC navn 2-[4-[2-[(4-chlorobenzoyl)amino]ethyl]phenoxy]-3,3,3-trideuterio-2-(trideuteriomethyl)propanoic acid
Molekylvægt (g/mol) 367.86
Formel vægt 367.1457