Learn More
Texwipe™ BetaMop™ II
Product Code. 11581214 Shop All Texwipe Products
Change view
Click to view available options
Product Type:
BetaMop II Mop
Mop Head Refill
For Use With (Equipment):
BetaMop II
Wring-bucket Systems
Unit Size:
Each
Pack of 6
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Product Type For Use With (Equipment) Unit Size
11581214 BetaMop II Mop Wring-bucket Systems Each
11541224 Mop Head Refill BetaMop II Pack of 6
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 11581214 Supplier Texwipe™ Supplier No. TX7106
  • ONLINE EXCLUSIVE PRICE
    1195.00 DKK
    1018.75 DKK / Pakke med 1
    Spare 176.25 DKK
    14% Slukket
Forventet forsendelse: 26-01-2027
Low contamination properties make this the perfect mop for floor cleaning in critical environments
Durable, absorbent, corrosion-resistant, and low-contaminating; Autoclavable.
Durability, corrosion resistance, and low contamination properties make this the perfect mop for floor cleaning in critical environments. High-capacity, seamless, tubular-knit polyester mop head provides excellent absorbency. BetaMop II may be used with all wring-bucket systems. Both the mop head and clamp are autoclavable for use in sterile environments.
Specifications
Product Type BetaMop II Mop
Autoclavable Autoclavable
Resistant To Corrosion
Includes Handle and Head Refill
For Use With (Equipment) Wring-bucket Systems
For Use With (Application) For floor cleaning in both electronic cleanrooms and sterile environments
Material Polyester