Få mere at vide
Betamethasone Tripropionate, TRC
Artikelnummer. 30011318
Skift visning
Mængde:
10 mg
25 mg
50 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30011318 10 mg
30009220 25 mg
30010351 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30011318 Leverandør Toronto Research Chemicals Leverandørnr. TRCB32705010MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 1186048-33-8
Molekylær formel C31 H41 F O8
Molekylvægt (g/mol) 560.65
Synonym (11beta,16beta)-9-Fluoro-16-methyl-11,17,21-tris(1-oxopropoxy)pregna-1,4-diene-3,20-dione, Betamethasone 11,17,21-tripropionate, 9-Fluoro-16beta-methyl-3,20-dioxopregna-1,4-diene-11beta,17,21-triyl Tripropanoate, Betamethasone Tripropionate, Betamethasone Dipropionate Imp. G (Pharmeuropa)
IUPAC navn [2-[(8S,9R,10S,11S,13S,14S,16S,17R)-9-fluoro-10,13,16-trimethyl-3-oxo-11,17-di(propanoyloxy)-6,7,8,11,12,14,15,16-octahydrocyclopenta[a]phenanthren-17-yl]-2-oxoethyl] propanoate
SMIL CCC(=O)OCC(=O)[C@@]1(OC(=O)CC)[C@@H](C)C[C@H]2[C@@H]3CCC4=CC(=O)C=C[C@]4(C)[C@@]3(F)[C@H](C[C@]12C)OC(=O)CC
Tekniske data
Kemisk navn eller materiale Betamethasone Tripropionate
Mængde 10 mg
InChI formel InChI=1S/C31H41FO8/c1-7-25(35)38-17-23(34)31(40-27(37)9-3)18(4)14-22-21-11-10-19-15-20(33)12-13-28(19,5)30(21,32)24(16-29(22,31)6)39-26(36)8-2/h12-13,15,18,21-22,24H,7-11,14,16-17H2,1-6H3/t18-,21-,22-,24-,28-,29-,30-,31-/m0/s1
Formel vægt 560.2785