Få mere at vide
Betamethasone Dipropionate, TRC
Artikelnummer. 30136348
Skift visning
Mængde:
100 mg
500 mg
1 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30136348 500 mg
30135288 1 g
17980655 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30136348 Leverandør Toronto Research Chemicals Leverandørnr. TRCB327005500MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 5593-20-4
Molekylær formel C28 H37 F O7
Molekylvægt (g/mol) 504.59
Synonym Pregna-1,4-diene-3,20-dione, 9-fluoro-11β,17,21-trihydroxy-16β-methyl-, 17,21-dipropionate (7CI,8CI), (11β,16β)-9-Fluoro-11-hydroxy-16-methyl-17,21-bis(1-oxopropoxy)pregna-1,4-diene-3,20-dione, Betamethasone 17,21-dipropionate, Betamethasone dipropionate, Diproderm, Diprolene, Diprophos, Diprosen, Diprosis, Diprosone, Diprospan, Maxivate, Psorion, Rinderon DP, S 3440, Sch 11460, β-Methasone 17,21-dipropionate
IUPAC navn [2-[(8S,9R,10S,11S,13S,14S,16S,17R)-9-fluoro-11-hydroxy-10,13,16-trimethyl-3-oxo-17-propanoyloxy-6,7,8,11,12,14,15,16-octahydrocyclopenta[a]phenanthren-17-yl]-2-oxoethyl] propanoate
SMIL CCC(=O)OCC(=O)[C@@]1(OC(=O)CC)[C@@H](C)C[C@H]2[C@@H]3CCC4=CC(=O)C=C[C@]4(C)[C@@]3(F)[C@@H](O)C[C@]12C
Tekniske data
Kemisk navn eller materiale Betamethasone-17,21-dipropionate
Analyt- eller komponentnavne Betamethasone-17,21-dipropionate
Forsendelsestilstand Room Temperature
InChI formel InChI=1S/C28H37FO7/c1-6-23(33)35-15-22(32)28(36-24(34)7-2)16(3)12-20-19-9-8-17-13-18(30)10-11-25(17,4)27(19,29)21(31)14-26(20,28)5/h10-11,13,16,19-21,31H,6-9,12,14-15H2,1-5H3/t16-,19-,20-,21-,25-,26-,27-,28-/m0/s1
Løsningstype Neat
Industry Type Pharmaceutical
Mængde 500 mg
Formel vægt 504.2523