Få mere at vide
Betamethasone Dipropionate, LoGiCal
Artikelnummer. 30463252
Skift visning
Mængde:
10 mg
Artikelnummer. Mængde
30463252 10 mg
1 options
Artikelnummer. 30463252 Leverandør LoGiCal Leverandørnr. LGCFOR0202.00
LoGiCal's certified reference materials as neats, native solutions and stable isotope labelled solutions, with expert documentation and support. From forensic to clinical labs, choose LoGiCal for confidence, compliance, and analytical excellence.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Betamethasone-17,21-dipropionate
CAS 5593-20-4
Analyt- eller komponentnavne Betamethasone-17,21-dipropionate
Forsendelsestilstand Room Temperature
Molekylær formel C28 H37 F O7
Certificeringer/compliance ISO 9001
Synonym Betamethasone Dipropionate, (11β,16β)-9-Fluoro-11-hydroxy-16-methyl-17,21-bis(1-oxopropoxy)-pregna-1,4-diene-3,20-dione, Betamethasone 17,21-Dipropionate, Diproderm, Diprolene, Diprophos, Diprosis, Diprosone, Diprospan, Maxivate, Psorion, Rinderon DP, S 3440, Sch 11460, β-Methasone 17,21-dipropionate;
InChI formel InChI=1S/C28H37FO7/c1-6-23(33)35-15-22(32)28(36-24(34)7-2)16(3)12-20-19-9-8-17-13-18(30)10-11-25(17,4)27(19,29)21(31)14-26(20,28)5/h10-11,13,16,19-21,31H,6-9,12,14-15H2,1-5H3/t16-,19-,20-,21-,25-,26-,27-,28-/m0/s1
Løsningstype Neat
SMIL CCC(=O)OCC(=O)[C@@]1(OC(=O)CC)[C@@H](C)C[C@H]2[C@@H]3CCC4=CC(=O)C=C[C@]4(C)[C@@]3(F)[C@@H](O)C[C@]12C
IUPAC navn [2-[(8S,9R,10S,11S,13S,14S,16S,17R)-9-fluoro-11-hydroxy-10,13,16-trimethyl-3-oxo-17-propanoyloxy-6,7,8,11,12,14,15,16-octahydrocyclopenta[a]phenanthren-17-yl]-2-oxoethyl] propanoate
Industry Type Clinical Testing
Molekylvægt (g/mol) 504.59
Mængde 10 mg
Formel vægt 504.2523 g/mol