Få mere at vide
Betahistine-d3 Dihydrochloride, TRC
Artikelnummer. 30329849
Skift visning
Mængde:
5 mg
Artikelnummer. Mængde
30329849 5 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30329849 Leverandør Toronto Research Chemicals Leverandørnr. TRCB3240025MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Betahistine-d3 Dihydrochloride
CAS 244094-72-2
Molekylær formel C8 2H3 H9 N2 . 2 Cl H
Mængde 5 mg
Synonym 2-Pyridineethanamine, N-(methyl-d3)-, dihydrochloride (9CI), Betahistine-d3 dihydrochloride, Betahistine D3 dihydrochloride (methyl D3)
InChI formel InChI=1S/C8H12N2.2ClH/c1-9-7-5-8-4-2-3-6-10-8;;/h2-4,6,9H,5,7H2,1H3;2*1H/i1D3;;
SMIL Cl.Cl.[2H]C([2H])([2H])NCCc1ccccn1
IUPAC navn 2-pyridin-2-yl-N-(trideuteriomethyl)ethanamine;dihydrochloride
Molekylvægt (g/mol) 212.13
Formel vægt 211.0722