missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ BetaBasic™ 8 HPLC Columns, 5μm Particle Size
Thermo ScientificTM BetaBasicTM 8 HPLC Columns are available in limited dimensions. Please contact Customer Service for more details.
4088.16 DKK - 5835.34 DKK
Specifications
Column Format | Analytical Column |
---|---|
For Use With | LC/MS, APCI |
Particle Size | 5 μm |
pH | 2 to 9 |
Pore Size | 150 Å |
Product Code | Brand | Particle Size | Diameter (Metric) | Length (Metric) | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Particle Size | Diameter (Metric) | Length (Metric) | Price | Quantity & Availability | |||
10417064
|
Thermo Scientific™
71405-052130 |
5 μm | 2.1 mm | 50 mm |
5723.71 DKK
Each |
In Stock
Log in to see stock. |
|||
10221095
|
Thermo Scientific™
71405-154630 |
5 μm | 4.6 mm | 150 mm |
5835.34 DKK
Each |
In Stock
Log in to see stock. |
|||
10240955
|
Thermo Scientific™
71405-054630 |
5 μm | 4.6 mm | 50 mm |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||
Description
Thermo ScientificTM BetaBasicTM 8 HPLC Columns are available in limited dimensions. Please contact Customer Service for more details.
- Pore size suitable for small molecules, peptides and protein digests
- Superb pH stability
Ordering Information:
Other column dimensions are also available. Please call Customer Service for more information.
Specifications
Analytical Column | |
5 μm | |
150 Å | |
L7 | |
Silica | |
Reversed Phase | |
Reversed Phase |
LC/MS, APCI | |
2 to 9 | |
200 m²/g | |
C8 | |
7% | |
Yes |