Få mere at vide
Beta-Zearalenol-d4 (Major), TRC
Artikelnummer. 30454924
Skift visning
Mængde:
10 mg
Artikelnummer. Mængde
30454924 10 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30454924 Leverandør Toronto Research Chemicals Leverandørnr. TRCZ27047810MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale beta-Zearalenol-d4
CAS 1778735-09-3
Renhedsgrad noter HPLC
Molekylær formel C18H20D4O5
Mængde 10 mg
Synonym (3S,7S,11E)-3,4,5,6,7,8,9,10-Octahydro-7,14,16-trihydroxy-3-methyl-1H-2-benzoxacyclotetradecin-1-one-d4, (-)-β-Zearalenol-d4, β-trans-Zearalenol-d4
InChI formel InChI=1S/C18H24O5/c1-12-6-5-9-14(19)8-4-2-3-7-13-10-15(20)11-16(21)17(13)18(22)23-12/h3,7,10-12,14,19-21H,2,4-6,8-9H2,1H3/b7-3+/t12-,14-/m0/s1/i8D2,9D2
SMIL O=C1O[C@@H](C)CCC([2H])([2H])[C@@H](O)C([2H])([2H])CC/C=C/C2=CC(O)=CC(O)=C21
IUPAC navn (2E,7S,11S)-6,6,8,8-tetradeuterio-7,15,17-trihydroxy-11-methyl-12-oxabicyclo[12.4.0]octadeca-1(18),2,14,16-tetraen-13-one
Molekylvægt (g/mol) 324.4
Formel vægt 324.1875
Procent renhed >95