Få mere at vide
Beta-Sitosterol-d7 (mixture of diasteromers), TRC
Artikelnummer. 30420700
Skift visning
Mængde:
100 mg
Artikelnummer. Mængde
30420700 100 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30420700 Leverandør Toronto Research Chemicals Leverandørnr. TRCS497052100MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale beta-Sitosterol-d7 (mixture of diasteromers)
CAS 2260669-28-9
Molekylær formel C29 D7 H43 O
Mængde 100 mg
Synonym 5-Cholesten-24(RS)-ethyl-3β-ol-25,26,26,26,27,27,27-d7, 24(R,S)-Stigmast-5-en-3β-ol
InChI formel InChI=1S/C29H50O/c1-7-21(19(2)3)9-8-20(4)25-12-13-26-24-11-10-22-18-23(30)14-16-28(22,5)27(24)15-17-29(25,26)6/h10,19-21,23-27,30H,7-9,11-18H2,1-6H3/t20-,21?,23+,24+,25-,26+,27+,28+,29-/m1/s1/i2D3,3D3,19D
SMIL [2H]C([2H])([2H])C([2H])(C(CC)CC[C@@H](C)[C@H]1CC[C@H]2[C@@H]3CC=C4C[C@@H](O)CC[C@]4(C)[C@H]3CC[C@]12C)C([2H])([2H])[2H]
IUPAC navn (3S,8S,9S,10R,13R,14S,17R)-10,13-dimethyl-17-[(2R)-6,7,7,7-tetradeuterio-5-ethyl-6-(trideuteriomethyl)heptan-2-yl]-2,3,4,7,8,9,11,12,14,15,16,17-dodecahydro-1H-cyclopenta[a]phenanthren-3-ol
Molekylvægt (g/mol) 421.75
Formel vægt 421.4301