missing translation for 'onlineSavingsMsg'
Få mere at vide

Beta-Sitosterol Beta-D-Glucoside, TRC

Artikelnummer. 30005897
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
5 mg
10 mg
25 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30005897 25 mg
30005756 10 mg
30008886 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 30005897 Leverandør Toronto Research Chemicals Leverandørnr. TRCS49701025MG

Please to purchase this item. Need a web account? Register with us today!

Denne vare kan ikke returneres. Se returpolicy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Kemiske identifikatorer

CAS 474-58-8
Molekylær formel C35 H60 O6
Molekylvægt (g/mol) 576.85
Synonym β-D-Glucopyranoside, (3β)-stigmast-5-en-3-yl, Daucosterin (6CI), Stigmast-5-ene, 3β-(β-D-glucopyranosyloxy)- (8CI), (3β)-Stigmast-5-en-3-yl β-D-glucopyranoside, Stigmastane, β-D-glucopyranoside deriv., (-)-β-Sitosterol glucoside, (-)-β-Sitosterol β-D-glucopyranoside, 1513DMIa, 3-(β-D-Glucopyranosyloxy)stigmast-5-ene, 3-O-D-Glucopyranosyl-β-sitosterol, 3-O-β-D-Glucopyranosyl-β-sitosterol, 3-O-β-D-Glucopyranosylsitosterol, 3-O-β-D-Glucosyl-β-sitosterol, 3β-D-Glucosyl-β-sitosterol, 3β-Sitoster-3-yl β-D-glucopyranoside, AW 10, Alexandrin, BSSG, Coriandrinol, Daucosterine, Daucosterol, Doursterol, EU 4906, Eleutheroside A, NSC 165962, Sitogluside, Sitosterol 3-O-β-D-glucopyranoside, Sitosterol 3-O-β-glucopyranoside, Sitosterol 3-O-β-glucoside, Sitosterol D-glucoside, Sitosterol glucopyranoside, Sitosterol glucoside, Sitosteryl 3β-D-glucopyranoside, Sitosteryl 3β-glucoside, Sitosteryl glucoside, Sitosteryl β-D-glucopyranoside, Sterolin, Stigmast-5-en-3-β-glucopyranoside, Stigmast-5-en-3β-yl β-D-glucopyranoside, Stigmast-5-ene, 3-(β-D-glucopyranosyloxy)-, (3β)-, Strumaroside, WA 184, β-Daucosterin, β-Daucosterol, β-Sitosterol 3-O-D-glucoside, β-Sitosterol 3-O-glucoside, β-Sitosterol 3-O-β-D-glucopyranoside, β-Sitosterol 3-O-β-D-glucoside, β-Sitosterol 3-O-β-glucopyranoside, β-Sitosterol 3-β-D-glucopyranoside, β-Sitosterol D-glucopyranoside, β-Sitosterol D-glucoside, β-Sitosterol O-glucoside, β-Sitosterol O-β-D-glucoside, β-Sitosterol glucopyranoside, β-Sitosterol glucoside, β-Sitosterol monoglucoside, β-Sitosterol β-D-glucopyranoside, β-Sitosterol β-D-glucoside, β-Sitosterol β-D-glycopyranoside, β-Sitosterol β-glucopyranoside, β-Sitosterol β-glucoside, β-Sitosterol-3-glucoside, β-Sitosteryl 3-O-glucoside, β-Sitosteryl 3-O-β-D-glucopyranoside, β-Sitosteryl 3-O-β-D-glucoside, β-Sitosteryl 3-glucoside, β-Sitosteryl 3-β-D-glucoside, β-Sitosteryl glucoside, β-Sitosteryl β-D-glucopyranoside, β-Sitosteryl β-D-glucoside
IUPAC navn (2R,3R,4S,5S,6R)-2-[[(3S,8S,9S,10R,13R,14S,17R)-17-[(2R,5R)-5-ethyl-6-methylheptan-2-yl]-10,13-dimethyl-2,3,4,7,8,9,11,12,14,15,16,17-dodecahydro-1H-cyclopenta[a]phenanthren-3-yl]oxy]-6-(hydroxymethyl)oxane-3,4,5-triol
SMIL CC[C@H](CC[C@@H](C)[C@H]1CC[C@H]2[C@@H]3CC=C4C[C@H](CC[C@]4(C)[C@H]3CC[C@]12C)O[C@@H]5O[C@H](CO)[C@@H](O)[C@H](O)[C@H]5O)C(C)C

Tekniske data

Kemisk navn eller materiale beta-Sitosterol-3-O-Glucoside
Mængde 25 mg
InChI formel InChI=1S/C35H60O6/c1-7-22(20(2)3)9-8-21(4)26-12-13-27-25-11-10-23-18-24(14-16-34(23,5)28(25)15-17-35(26,27)6)40-33-32(39)31(38)30(37)29(19-36)41-33/h10,20-22,24-33,36-39H,7-9,11-19H2,1-6H3/t21-,22-,24+,25+,26-,27+,28+,29-,30-,31+,32-,33-,34+,35-/m1/s1
Formel vægt 576.439
Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.