missing translation for 'onlineSavingsMsg'
Få mere at vide

Beta-Phenylethyl Beta-D-Glucoside Tetraacetate, TRC

Artikelnummer. 30417614
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
500 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30417614 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 30417614 Leverandør Toronto Research Chemicals Leverandørnr. TRCP320870500MG

Please to purchase this item. Need a web account? Register with us today!

Denne vare kan ikke returneres. Se returpolicy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Kemisk navn eller materiale beta-Phenylethyl beta-D-Glucoside Tetraacetate
CAS 76870-87-6
Molekylær formel C22H28O10
Mængde 500 mg
Synonym beta-Phenylethyl beta-D-Glucoside 2,3,4,6-Tetraacetate, 2-Phenylethyl-2,3,4,6-tetra-O-acetyl-beta-D-glucopyranoside, NSC 231829
InChI formel InChI=1S/C22H28O10/c1-13(23)28-12-18-19(29-14(2)24)20(30-15(3)25)21(31-16(4)26)22(32-18)27-11-10-17-8-6-5-7-9-17/h5-9,18-22H,10-12H2,1-4H3
SMIL CC(=O)OCC1OC(OCCc2ccccc2)C(OC(=O)C)C(OC(=O)C)C1OC(=O)C
IUPAC navn [3,4,5-triacetyloxy-6-(2-phenylethoxy)oxan-2-yl]methyl acetate
Molekylvægt (g/mol) 452.45
Formel vægt 452.1682
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.