Få mere at vide
Beta-(p-Nitrophenyl)ethyl Bromide-d4, TRC
Artikelnummer. 30472390
Skift visning
Mængde:
100 mg
Artikelnummer. Mængde
30472390 100 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30472390 Leverandør Toronto Research Chemicals Leverandørnr. TRCN503432100MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale beta-(p-Nitrophenyl)ethyl Bromide-d4
CAS 1794971-35-9
Molekylær formel C8H4D4BrNO2
Mængde 100 mg
Synonym 1-(2-Bromoethyl)-4-nitrobenzene-d4, 1-Bromo-2-(4-nitrophenyl)ethane-d4, 2-(4-Nitrophenyl)-1-bromoethane, -d4 2-(4-Nitrophenyl)ethyl Bromide-d4, 2-(p-Nitrophenyl)ethyl Bromide-d4, 4-(2-Bromoethyl)-1-nitrobenzene-d4, 4-(2-Bromoethyl)nitrobenzene-d4, 4-Nitrophenethyl Bromide-d4, NSC 100730-d4, NSC 3493-d4, p-Nitrophenethyl Bromide-d4, β-(p-Nitrophenyl)ethyl Bromide-d4
InChI formel InChI=1S/C8H8BrNO2/c9-6-5-7-1-3-8(4-2-7)10(11)12/h1-4H,5-6H2/i5D2,6D2
SMIL BrC([2H])([2H])C([2H])([2H])C1=CC=C([N+]([O-])=O)C=C1
Molekylvægt (g/mol) 234.08
Formel vægt 232.9989