missing translation for 'onlineSavingsMsg'
Learn More
Learn More
beta-Nicotinamide adenine dinucleotide phosphate reduced tetrasodium salt hydrate, 97%, Thermo Scientific Chemicals
Click to view available options
Quantity:
100 mg
500 mg
Description
beta-Nicotinamide adenine dinucleotide phosphate reduced tetrasodium salt hydrate is used as NADP+/NADPH redox pair is used in a variety of antioxidation mechanism where it protects agains reactive oxidation species accumulation. NADPH is generated in vivio by the pentose phosphate pathway.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
beta-Nicotinamide adenine dinucleotide phosphate reduced tetrasodium salt hydrate is used as NADP+/NADPH redox pair is used in a variety of antioxidation mechanism where it protects agains reactive oxidation species accumulation. NADPH is generated in vivio by the pentose phosphate pathway.
Solubility
It is soluble in 10 mM NaOH.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Recommended storage temperature -20°C Light sensitive. Hygroscopic
beta-Nicotinamide adenine dinucleotide phosphate reduced tetrasodium salt hydrate is used as NADP+/NADPH redox pair is used in a variety of antioxidation mechanism where it protects agains reactive oxidation species accumulation. NADPH is generated in vivio by the pentose phosphate pathway.
Solubility
It is soluble in 10 mM NaOH.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Recommended storage temperature -20°C Light sensitive. Hygroscopic
Specifications
Color | White |
Quantity | 100 mg |
Solubility Information | It is soluble in 10 mM NaOH. |
Formula Weight | 905.42 (833.35 Anhydrous) |
Percent Purity | ≥98% |
Physical Form | Powder |
Chemical Name or Material | beta-Nicotinamide adenine dinucleotide phosphate reduced tetrasodium salt hydrate |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'