missing translation for 'onlineSavingsMsg'
Learn More

beta-Nicotinamide adenine dinucleotide phosphate reduced tetrasodium salt hydrate, 97%, Thermo Scientific Chemicals

Catalog Number 15434455
Click to view available options
Quantity:
100 mg
500 mg
C21H26N7Na4O17P3·xH2O
MFCD07366294
beta-NADPH tetrasodium salt tetrahydrate
C21H26N7Na4O17P3·xH2O
MFCD07366294
beta-NADPH tetrasodium salt tetrahydrate

beta-Nicotinamide adenine dinucleotide phosphate reduced tetrasodium salt hydrate is used as NADP+/NADPH redox pair is used in a variety of antioxidation mechanism where it protects agains reactive oxidation species accumulation. NADPH is generated in vivio by the pentose phosphate pathway.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
beta-Nicotinamide adenine dinucleotide phosphate reduced tetrasodium salt hydrate is used as NADP+/NADPH redox pair is used in a variety of antioxidation mechanism where it protects agains reactive oxidation species accumulation. NADPH is generated in vivio by the pentose phosphate pathway.

Solubility
It is soluble in 10 mM NaOH.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Recommended storage temperature -20°C Light sensitive. Hygroscopic
TRUSTED_SUSTAINABILITY
Color White
Quantity 100 mg
Solubility Information It is soluble in 10 mM NaOH.
Formula Weight 905.42 (833.35 Anhydrous)
Percent Purity ≥98%
Physical Form Powder
Chemical Name or Material beta-Nicotinamide adenine dinucleotide phosphate reduced tetrasodium salt hydrate

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'