missing translation for 'onlineSavingsMsg'
Learn More

beta-Nicotinamide adenine dinucleotide monosodium salt dihydrate, Thermo Scientific™

Catalog Number 15865468
Click to view available options
Quantity:
5 g
100 g
200 g
Packaging:
Plastic bottle
This item is not returnable. View return policy
20111-18-6
C21H26N7NaO14P2
685.41
MFCD30724412
OGCURMAMSJFXSG-QYZPTAICSA-M
adenosine5'-trihydrogen diphosphate , p'
131875671
[[(2R,3S,4R,5R)-5-(6-aminopurin-9-yl)-3,4-dihydroxyoxolan-2-yl]methoxy-hydroxyphosphoryl] [(2R,3S,4R,5R)-5-(3-carbamoylpyridin-1-ium-1-yl)-3,4-dihydroxyoxolan-2-yl]methyl phosphate;sodium
[Na+].NC(=O)C1=CC=C[N+](=C1)[C@@H]1O[C@H](COP([O-])(=O)OP([O-])(=O)OC[C@H]2O[C@H]([C@H](O)[C@@H]2O)N2C=NC3=C(N)N=CN=C23)[C@@H](O)[C@H]1O
This item is not returnable. View return policy
20111-18-6
C21H26N7NaO14P2
685.41
MFCD30724412
OGCURMAMSJFXSG-QYZPTAICSA-M
adenosine5'-trihydrogen diphosphate , p'
131875671
[[(2R,3S,4R,5R)-5-(6-aminopurin-9-yl)-3,4-dihydroxyoxolan-2-yl]methoxy-hydroxyphosphoryl] [(2R,3S,4R,5R)-5-(3-carbamoylpyridin-1-ium-1-yl)-3,4-dihydroxyoxolan-2-yl]methyl phosphate;sodium
[Na+].NC(=O)C1=CC=C[N+](=C1)[C@@H]1O[C@H](COP([O-])(=O)OP([O-])(=O)OC[C@H]2O[C@H]([C@H](O)[C@@H]2O)N2C=NC3=C(N)N=CN=C23)[C@@H](O)[C@H]1O

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Physical Form Powder
Quantity 200 g
Formula Weight 721.44
Packaging Plastic bottle
Chemical Name or Material beta-Nicotinamide adenine dinucleotide monosodium salt dihydrate
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'