missing translation for 'onlineSavingsMsg'
Få mere at vide
Få mere at vide
Beta-Naphthyl Beta-D-Glucopyranoside Tetraacetate, TRC
Specifications
Specifications
| Kemisk navn eller materiale | beta-Naphthyl beta-D-Glucopyranoside Tetraacetate |
| CAS | 14581-89-6 |
| Molekylær formel | C24 H26 O10 |
| Mængde | 100 mg |
| InChI formel | InChI=1S/C24H26O10/c1-13(25)29-12-20-21(30-14(2)26)22(31-15(3)27)23(32-16(4)28)24(34-20)33-19-10-9-17-7-5-6-8-18(17)11-19/h5-11,20-24H,12H2,1-4H3/t20-,21-,22+,23-,24-/m1/s1 |
| SMIL | CC(=O)OC[C@H]1O[C@@H](Oc2ccc3ccccc3c2)[C@H](OC(=O)C)[C@@H](OC(=O)C)[C@@H]1OC(=O)C |
| IUPAC navn | [(2R,3R,4S,5R,6S)-3,4,5-triacetyloxy-6-naphthalen-2-yloxyoxan-2-yl]methyl acetate |
| Molekylvægt (g/mol) | 474.46 |
| Formel vægt | 474.1526 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?