missing translation for 'onlineSavingsMsg'
Få mere at vide

Beta-Naphthalenesulfonyl-D-valine, TRC

Product Code. 30476548
Change view
Click to view available options
Mængde:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Mængde
30476548 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30476548 Supplier Toronto Research Chemicals Supplier No. TRCN359260100MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Kemisk navn eller materiale beta-Naphthalenesulfonyl-D-valine
CAS 182227-17-4
Molekylær formel C15H17N4S
Mængde 100 mg
Synonym N-(2-Naphthalenylsulfonyl)-D-valine
InChI formel InChI=1S/C15H17NO4S/c1-10(2)14(15(17)18)16-21(19,20)13-8-7-11-5-3-4-6-12(11)9-13/h3-10,14,16H,1-2H3,(H,17,18)/t14-/m1/s1
SMIL CC(C)[C@@H](NS(=O)(=O)c1ccc2ccccc2c1)C(=O)O
IUPAC navn (2R)-3-methyl-2-(naphthalen-2-ylsulfonylamino)butanoic acid
Molekylvægt (g/mol) 307.36
Formel vægt 307.0878
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.