Få mere at vide
Beta-Myrcene-d6, TRC
Artikelnummer. 30476836
Skift visning
Mængde:
2.5 mg
25 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30476836 25 mg
30475041 2.5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30476836 Leverandør Toronto Research Chemicals Leverandørnr. TRCM87530225MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale beta-Myrcene-d6
CAS 75351-99-4
Molekylær formel C10 D6 H10
Mængde 25 mg
Synonym 7-Methyl-3-methylene-1,6-octadiene-d6, 2-Methyl-6-methylene-2,7-octadiene-d6, 3-Methylene-7-methyl-1,6-octadiene-d6, Myrcene-d6, NSC 406264-d6, β-Geraniolene-d6
InChI formel InChI=1S/C10H16/c1-5-10(4)8-6-7-9(2)3/h5,7H,1,4,6,8H2,2-3H3/i2D3,3D3
SMIL [2H]C([2H])([2H])C(=CCCC(=C)C=C)C([2H])([2H])[2H]
IUPAC navn 8,8,8-trideuterio-3-methylene-7-(trideuteriomethyl)octa-1,6-diene
Molekylvægt (g/mol) 142.27
Formel vægt 142.163