Få mere at vide
Beta-Methylamino-L-alanine Dihydrochloride, TRC
Artikelnummer. 30475803
Skift visning
Mængde:
2.5 mg
Artikelnummer. Mængde
30475803 2.5 mg
1 options
Artikelnummer. 30475803 Leverandør Toronto Research Chemicals Leverandørnr. TRCM2855502.5MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale beta-Methylamino-L-alanine
CAS 2340337-60-0
Molekylær formel C4H10N2O2 .2(HCl)
Mængde 2.5 mg
Synonym 2-Amino-3-(methylamino)propanoic Acid Dihydrochloride, S(+)-β-Methyl-α,β-diaminopropionic Acid Dihydrochloride, BMAA Dihydrochloride
InChI formel InChI=1S/C4H10N2O2.2ClH/c1-6-2-3(5)4(7)8;;/h3,6H,2,5H2,1H3,(H,7,8);2*1H/t3-;;/m0../s1
SMIL N[C@@H](CNC)C(O)=O.[H]Cl.[H]Cl
IUPAC navn (S)-2-amino-3-(methylamino)propanoic acid dihydrochloride
Molekylvægt (g/mol) 191.05
Formel vægt 190.03