Få mere at vide
beta-(m-Hydroxyphenyl)hydracrylic Acid, TRC
Artikelnummer. 30136390
Skift visning
Mængde:
5 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30136390 50 mg
30005858 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30136390 Leverandør Toronto Research Chemicals Leverandørnr. TRCH94928050MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 3247-75-4
Molekylær formel C9 H10 O4
Molekylvægt (g/mol) 182.17
Synonym Benzenepropanoic acid, β,3-dihydroxy-, Hydracrylic acid, 3-(m-hydroxyphenyl)- (6CI,7CI,8CI), β,3-Dihydroxybenzenepropanoic acid, (3-Hydroxyphenyl)hydracrylic acid, 3-(3-Hydroxyphenyl)-3-hydroxypropionic acid, 3-(3-Hydroxyphenyl)hydracrylic acid, 3-(m-Hydroxyphenyl)hydracrylic acid, 3-Hydroxy-3-(3-hydroxyphenyl)propanoic acid, β-(m-Hydroxyphenyl)hydracrylic acid
IUPAC navn 3-hydroxy-3-(3-hydroxyphenyl)propanoic acid
SMIL OC(CC(=O)O)c1cccc(O)c1
Tekniske data
Kemisk navn eller materiale beta-(m-Hydroxyphenyl)hydracrylic Acid
Mængde 50 mg
InChI formel InChI=1S/C9H10O4/c10-7-3-1-2-6(4-7)8(11)5-9(12)13/h1-4,8,10-11H,5H2,(H,12,13)
Formel vægt 182.0579