missing translation for 'onlineSavingsMsg'
Få mere at vide

beta-Hydroxy Tamoxifen, TRC

Artikelnummer. 30406200
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
50 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30406200 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denne vare kan ikke returneres. Se returpolicy
Regulatorisk: Dette produkt falder ind under Lov om kemiske stoffer og produkter - lovbekendtgørelse nr. 1755 af 22. december 2006. Hvis det er relevant, skal kunden fremskaffe en licens før levering.
Artikelnummer. 30406200 Leverandør Toronto Research Chemicals Leverandørnr. TRCH95471550MG

Please to purchase this item. Need a web account? Register with us today!

Denne vare kan ikke returneres. Se returpolicy
Regulatorisk: Dette produkt falder ind under Lov om kemiske stoffer og produkter - lovbekendtgørelse nr. 1755 af 22. december 2006. Hvis det er relevant, skal kunden fremskaffe en licens før levering.

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Tekniske data

Kemisk navn eller materiale beta-Hydroxy Tamoxifen
CAS 97151-03-6
Molekylær formel C26 H29 N O2
Mængde 50 mg
Synonym Benzenepropanol, γ-[[4-[2-(dimethylamino)ethoxy]phenyl]phenylmethylene]-, (γZ)-, Benzenepropanol, γ-[[4-[2-(dimethylamino)ethoxy]phenyl]phenylmethylene]-, (Z)-, (γZ)-γ-[[4-[2-(Dimethylamino)ethoxy]phenyl]phenylmethylene]benzenepropanol, Tamoxifen alcohol, β-Hydroxytamoxifen
InChI formel InChI=1S/C26H29NO2/c1-27(2)18-20-29-24-15-13-23(14-16-24)26(22-11-7-4-8-12-22)25(17-19-28)21-9-5-3-6-10-21/h3-16,28H,17-20H2,1-2H3/b26-25-
SMIL CN(C)CCOc1ccc(cc1)\C(=C(\CCO)/c2ccccc2)\c3ccccc3
IUPAC navn (Z)-4-[4-[2-(dimethylamino)ethoxy]phenyl]-3,4-diphenylbut-3-en-1-ol
Molekylvægt (g/mol) 387.51
Formel vægt 387.2198
Produkttitel
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.