Få mere at vide
Beta-Guaiene (>85%), TRC
Artikelnummer. 30180423
Skift visning
Mængde:
10 mg
50 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30180423 50 mg
30180320 10 mg
30179958 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30180423 Leverandør Toronto Research Chemicals Leverandørnr. TRCG81100550MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 88-84-6
Molekylær formel C15H24
Molekylvægt (g/mol) 204.35
Synonym (1S,4S)-1,4-Dimethyl-7-(propan-2-ylidene)-1,2,3,4,5,6,7,8-octahydroazulene;(1S,4S)-1,2,3,4,5,6,7,8-Octahydro-1,4-dimethyl-7-(1-methylethylidene)azulene;(1S-cis)-1,2,3,4,5,6,7,8-octahydro-1,4-dimethyl-7-(1-methylethylidene)-Azulene;Guaia-1(5),7(11)-diene;Guaiene;beta-Guajene;
IUPAC navn (1S,4S)-1,4-dimethyl-7-propan-2-ylidene-2,3,4,5,6,8-hexahydro-1H-azulene
SMIL C[C@H]1CCC(=C(C)C)CC2=C1CC[C@@H]2C
Tekniske data
Kemisk navn eller materiale beta-Guaiene
Mængde 50 mg
Formel vægt 204.1878