missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ beta-D-Gentiobiose, 98%

Brand:  Alfa Aesar™ J66750.03

Additional Details : CAS Number : 554-91-6 Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15438229

  • 2056.00 DKK / 1g

Estimated Shipment
08-04-2021

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Applications
Gentiobiose was used in binding assays to determine if coelomic cytolytic factor 1 (CCF-1) binds to β-1,3-glucans to stimulate the pro-PO-activating system.

Solubility
Slightly soluble in water.

Notes
Store in cool, dry conditions in well sealed containers. Keep container tightly closed.
Specifications

Chemical Identifiers

554-91-6
C12H22O11
MFCD00198056
6-O-(beta-D-Glucopyranosyl)-D-glucopyranose; Amygdalose

Specifications

beta-D-Gentiobiose
1g
C12H22O11
93354
Slightly soluble in water.
Powder
White
554-91-6
MFCD00198056
6-O-(beta-D-Glucopyranosyl)-D-glucopyranose; Amygdalose
342.3
98%
Videos
SDS Documents