Få mere at vide
Beta-Cyclogeranyltriphenylphosphonium Bromide-d5, TRC
Artikelnummer. 30362001
Skift visning
Mængde:
50 mg
Artikelnummer. Mængde
30362001 50 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30362001 Leverandør Toronto Research Chemicals Leverandørnr. TRCC98785250MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale beta-Cyclogeranyltriphenylphosphonium Bromide-d5
CAS 78995-97-8
Molekylær formel C28H27D5BrP
Mængde 50 mg
Synonym Triphenyl[(2,6,6-trimethyl-1-cyclohexen-1-yl)methyl]phosphonium Bromide-d5
InChI formel InChI=1S/C28H32P.BrH/c1-23-14-13-21-28(2,3)27(23)22-29(24-15-7-4-8-16-24,25-17-9-5-10-18-25)26-19-11-6-12-20-26;/h4-12,15-20H,13-14,21-22H2,1-3H3;1H/q+1;/p-1/i1D3,14D2;
SMIL [Br-].[2H]C([2H])([2H])C1=C(C[P+](c2ccccc2)(c3ccccc3)c4ccccc4)C(C)(C)CCC1([2H])[2H]
IUPAC navn [3,3-dideuterio-6,6-dimethyl-2-(trideuteriomethyl)cyclohexen-1-yl]methyl-triphenylphosphanium;bromide
Molekylvægt (g/mol) 484.46
Formel vægt 483.1739