Få mere at vide
Beta-Cyano-Beta-methyl-hydrocinnamic Acid Ethyl Ester, TRC
Artikelnummer. 30351158
Skift visning
Mængde:
5 g
Artikelnummer. Mængde
30351158 5 g
1 options
Artikelnummer. 30351158 Leverandør Toronto Research Chemicals Leverandørnr. TRCC9819705G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale beta-Cyano-beta-methyl-hydrocinnamic Acid Ethyl Ester
CAS 123064-05-1
Molekylær formel C13H15NO2
Mængde 5 g
Synonym beta-Cyano-beta-methyl-benzenepropanoic Acid Ethyl Ester
InChI formel InChI=1S/C13H15NO2/c1-3-16-12(15)9-13(2,10-14)11-7-5-4-6-8-11/h4-8H,3,9H2,1-2H3
SMIL CCOC(=O)CC(C)(C#N)c1ccccc1
IUPAC navn ethyl 3-cyano-3-phenylbutanoate
Molekylvægt (g/mol) 217.26
Formel vægt 217.1103