Få mere at vide
beta-Costol, TRC
Artikelnummer. 30010862
Skift visning
Mængde:
0.5 mg
2.5 mg
5 mg
Pakningsstørrelse:
2.5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
30010862 2.5 mg 2.5mg
30009584 0.5 mg -
30009447 5 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30010862 Leverandør Toronto Research Chemicals Leverandørnr. TRCC6975702.5MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 515-20-8
Molekylær formel C15 H24 O
Molekylvægt (g/mol) 220.351
Synonym [2R-(2α,4aα,8aβ)]-Decahydro-4a-methyl-β,8-bis(methylene)-2-Naphthaleneethanol;2-((2R,4aR,8aS)-4a-Methyl-8-methylenedecahydronaphthalen-2-yl)prop-2-en-1-ol;(2R,4aR,8aS)-Decahydro-4a-methyl-β,8-bis(methylene)-2-naphthaleneethanol;(+)-Costol;(+)-β-Costol;Sesquibenihiol;Costol;Eudesma-4(14),11(13)-dien-12-ol;
IUPAC navn 2-[(2R,4aR,8aS)-4a-methyl-8-methylidene-1,2,3,4,5,6,7,8a-octahydronaphthalen-2-yl]prop-2-en-1-ol
SMIL C[C@]12CCCC(=C)[C@@H]1C[C@@H](CC2)C(=C)CO
Tekniske data
Kemisk navn eller materiale beta-Costol
Mængde 2.5 mg
InChI formel InChI=1S/C15H24O/c1-11-5-4-7-15(3)8-6-13(9-14(11)15)12(2)10-16/h13-14,16H,1-2,4-10H2,3H3/t13-,14+,15-/m1/s1
Formel vægt 220.183