Få mere at vide
Beta-Cortolone, TRC
Artikelnummer. 30133635
Skift visning
Mængde:
10 mg
25 mg
50 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30133635 25 mg
17923195 50 mg
17914415 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30133635 Leverandør Toronto Research Chemicals Leverandørnr. TRCC69628525MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 667-66-3
Molekylær formel C21 H34 O5
Molekylvægt (g/mol) 366.49
Synonym 3α,17,20β,21-Tetrahydroxy-11-pregnanone, 5β-Pregnan-11-one, 3α,17,20β,21-tetrahydroxy- (7CI,8CI), 20β-Cortolone, 20β-Hexahydrocortisone, 3α,17,20β,21-Tetrahydroxy-5β-pregnan-11-one, 3α,17α,20β,21-Tetrahydroxy-5β-pregnan-11-one, 5β-Pregnan-3α,17α,20β,21-tetrol-11-one, 5β-Pregnane-3α,17α,20β,21-tetrol-11-one, NSC 56997
IUPAC navn (3R,5R,8S,9S,10S,13S,14S,17R)-17-[(1R)-1,2-dihydroxyethyl]-3,17-dihydroxy-10,13-dimethyl-2,3,4,5,6,7,8,9,12,14,15,16-dodecahydro-1H-cyclopenta[a]phenanthren-11-one
SMIL C[C@]12CC[C@@H](O)C[C@H]1CC[C@H]3[C@@H]4CC[C@](O)([C@H](O)CO)[C@@]4(C)CC(=O)[C@H]23
Tekniske data
Kemisk navn eller materiale beta-Cortolone
Mængde 25 mg
InChI formel InChI=1S/C21H34O5/c1-19-7-5-13(23)9-12(19)3-4-14-15-6-8-21(26,17(25)11-22)20(15,2)10-16(24)18(14)19/h12-15,17-18,22-23,25-26H,3-11H2,1-2H3/t12-,13-,14+,15+,17-,18-,19+,20+,21+/m1/s1
Formel vægt 366.2406